LIF509446793 Life Chemicals Inc.

5-((methylamino)methyl)pyridin-2(1H)-one dihydrobromide

  • MDL number MFCD30011202
  • Cas Number : 1949815-78-4
  • Molecular Weight : 299.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart