LIF509446795 Life Chemicals Inc.

1-(6-methoxypyridin-3-yl)-N-methylmethanamine dihydrochloride

  • MDL number MFCD30011204
  • Cas Number : 1949816-05-0
  • Molecular Weight : 225.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart