LIF509446797 Life Chemicals Inc.

6-(dimethylamino)nicotinic acid hydrochloride

  • MDL number MFCD30011206
  • Cas Number : 1949815-89-7
  • Molecular Weight : 202.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart