LIF509446849 Life Chemicals Inc.

2-[1-(2-methylpropyl)-1H-pyrazol-5-yl]cyclopropane-1-carboxylic acid

  • MDL number MFCD28133326
  • Cas Number : 1824055-05-1
  • Molecular Weight : 208.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart