LIF509446851 Life Chemicals Inc.

2-[1-(propan-2-yl)-1H-pyrazol-5-yl]cyclopropane-1-carboxylic acid

  • MDL number MFCD28133328
  • Cas Number : 1823322-75-3
  • Molecular Weight : 194.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart