LIF509446857 Life Chemicals Inc.

tert-butyl 3-bromo-2-oxopyrrolidine-1-carboxylate

  • MDL number MFCD28133340
  • Cas Number : 1824027-05-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart