LIF509446885 Life Chemicals Inc.

1-ethyl-1H-pyrrole-2-carbaldehyde

  • MDL number MFCD11909543
  • Cas Number : 2167-14-8
  • Molecular Weight : 123.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart