LIF509446898 Life Chemicals Inc.

6-cyclopropyl-2,3-dihydropyridazin-3-one

  • MDL number MFCD16871334
  • Cas Number : 1161737-33-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart