LIF509446904 Life Chemicals Inc.

5-phenylpent-1-yn-3-ol

  • MDL number MFCD20942935
  • Cas Number : 67648-04-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart