LIF509446906 Life Chemicals Inc.

5-bromo-2-methoxy-4-methylbenzaldehyde

  • MDL number MFCD21234728
  • Cas Number : 923281-67-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart