LIF509446910 Life Chemicals Inc.

2,2,2-trifluoro-1-(pyridin-4-yl)ethan-1-ol

  • MDL number MFCD16618051
  • Cas Number : 394203-58-8
  • Molecular Weight : 177.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart