LIF509446911 Life Chemicals Inc.

2-[(prop-2-yn-1-yl)amino]acetic acid hydrochloride

  • MDL number MFCD29046441
  • Cas Number : 104501-33-9
  • Molecular Weight : 149.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart