LIF509446913 Life Chemicals Inc.

2-(3-methylphenyl)-1H-imidazole

  • MDL number MFCD12818507
  • Cas Number : 21202-37-9
  • Molecular Weight : 158.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart