LIF509446916 Life Chemicals Inc.

3-(1H-imidazol-1-yl)cyclohexan-1-amine

  • MDL number MFCD23734648
  • Cas Number : 1497207-17-6
  • Molecular Weight : 165.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart