LIF509446925 Life Chemicals Inc.

3-aminopentan-2-ol

  • MDL number MFCD11213645
  • Cas Number : 50411-28-4
  • Molecular Weight : 103.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart