LIF509446935 Life Chemicals Inc.

2-amino-1-(2,6-difluorophenyl)ethan-1-one hydrochloride

  • MDL number MFCD08741372
  • Cas Number : 1210255-19-8
  • Molecular Weight : 207.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart