LIF509446956 Life Chemicals Inc.

(6-chloro-1,3-benzothiazol-2-yl)methanamine

  • MDL number MFCD11902165
  • Cas Number : 50739-36-1
  • Molecular Weight : 198.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart