LIF509446975 Life Chemicals Inc.

(2-phenylcyclopropyl)methanamine hydrochloride

  • MDL number MFCD18483297
  • Cas Number : 90874-42-3
  • Molecular Weight : 183.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart