LIF509446976 Life Chemicals Inc.

methyl 2-(cyclopropylamino)acetate hydrochloride

  • MDL number MFCD22196409
  • Cas Number : 182291-97-0
  • Molecular Weight : 165.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart