LIF509446986 Life Chemicals Inc.

6-bromo-[1,3]oxazolo[5,4-b]pyridine

  • MDL number MFCD07375014
  • Cas Number : 886372-90-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart