LIF509447019 Life Chemicals Inc.

2-(1H-1,2,4-triazol-1-yl)propanoic acid

  • MDL number MFCD08361621
  • Cas Number : 901586-50-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart