LIF509447087 Life Chemicals Inc.

1-(2-bromoethyl)pyrrolidin-2-one

  • MDL number MFCD09804536
  • Cas Number : 117018-99-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart