LIF509447503 Life Chemicals Inc.

3-(4-fluorophenyl)-5H,6H,7H,8H,9H-imidazo[1,5-a]azepine-1-carboxylic acid

  • MDL number MFCD10686866
  • Cas Number : 1050886-00-4
  • Molecular Weight : 274.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart