LIF509447528 Life Chemicals Inc.

4-methyl-2-(methylsulfonyl)aniline

  • MDL number MFCD20699519
  • Cas Number : 4284-63-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart