LIF509447529 Life Chemicals Inc.

1-[(tert-butoxy)carbonyl]-3,3,5,5-tetramethylpiperidine-4-carboxylic acid

  • MDL number MFCD29090765
  • Cas Number : 1780565-88-9
  • Molecular Weight : 285.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart