LIF509447530 Life Chemicals Inc.

tert-butyl 3,3,5,5-tetramethyl-4-oxopiperidine-1-carboxylate

  • MDL number MFCD11975582
  • Cas Number : 1235553-77-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart