LIF509447532 Life Chemicals Inc.

1-(aminomethyl)-4-chloro-2,3-dihydroinden-1-ol

  • MDL number MFCD11591023
  • Cas Number : 1082563-13-0
  • Molecular Weight : 197.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart