LIF509447537 Life Chemicals Inc.

5-cyclohexyloxazole-4-carboxylic acid

  • MDL number MFCD07376164
  • Cas Number : 1226165-87-2
  • Molecular Weight : 195.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart