LIF509447541 Life Chemicals Inc.

5-chloro-2-(2,2,2-trifluoroethoxy)benzene-1-sulfonyl chloride

  • MDL number MFCD17301616
  • Cas Number : 111762-22-2
  • Molecular Weight : 309.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart