LIF509447587 Life Chemicals Inc.

1-(1H-indol-5-yl)ethan-1-one

  • MDL number MFCD00481875
  • Cas Number : 53330-94-2
  • Molecular Weight : 159.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart