LIF509447588 Life Chemicals Inc.

N-methyl-1-(5-phenyl-1,3,4-oxadiazol-2-yl)methanamine hydrochloride

  • MDL number MFCD06738853
  • Cas Number : 1158371-89-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart