LIF514346930 Life Chemicals Inc.

sodium 1-hydroxy-3-methylbutane-1-sulfonate hydrate

  • MDL number MFCD30342701

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart