LIF514346973 Life Chemicals Inc.

4-(4-methoxyphenyl)-6-(piperazin-1-yl)pyrimidine

  • MDL number MFCD21980463
  • Cas Number : 1368729-14-9
  • Molecular Weight : 270.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart