LIF514346986 Life Chemicals Inc.

ethyl 6-(hydroxymethyl)pyridine-3-carboxylate

  • MDL number MFCD12131130
  • Cas Number : 35005-81-3
  • Molecular Weight : 181.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart