LIF514347012 Life Chemicals Inc.

2-(2-fluorophenyl)-2-methoxypropan-1-amine

  • MDL number MFCD28369948
  • Cas Number : 933751-44-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart