LIF514347114 Life Chemicals Inc.

2-methylbenzimidazole-5-carboxylic acid

  • MDL number MFCD00463064
  • Cas Number : 709-19-3
  • Molecular Weight : 176.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart