LIF514347210 Life Chemicals Inc.

3-(2-chloroethyl)-5-methyl-1,2-oxazole

  • MDL number MFCD19234148
  • Cas Number : 14775-98-5
  • Molecular Weight : 145.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart