LIF514347222 Life Chemicals Inc.

4-(6-oxo-1,6-dihydropyridazin-1-yl)butanoic acid

  • MDL number MFCD17271176
  • Cas Number : 346407-15-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart