LIF514347223 Life Chemicals Inc.

4-chloro-N-ethylpyrimidin-2-amine

  • MDL number MFCD11046855
  • Cas Number : 86575-65-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart