LIF514347239 Life Chemicals Inc.

4,5,6,7-tetrahydro-2,1-benzoxazole-3-carboxylic acid

  • MDL number MFCD10566736
  • Cas Number : 261350-47-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart