LIF514347240 Life Chemicals Inc.

6-(morpholin-4-yl)pyridazin-3-amine

  • MDL number MFCD09743734
  • Cas Number : 66346-91-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart