LIF514347246 Life Chemicals Inc.

3-bromo-5-methoxypyridin-4-amine

  • MDL number MFCD28669603
  • Cas Number : 1417618-49-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart