LIF514347248 Life Chemicals Inc.

2-methyl-1H,2H,3H,4H,9H-pyrido[3,4-b]indole

  • MDL number MFCD00229120
  • Cas Number : 13100-00-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart