LIF517189467 Life Chemicals Inc.

5-bromo-3-iodo-4-methylpyridin-2-ol

  • MDL number MFCD30484393
  • Cas Number : 2031260-86-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart