LIF517189482 Life Chemicals Inc.

5-methoxy-4-methylpyridin-2-amine

  • MDL number MFCD28099901
  • Cas Number : 1800035-89-5
  • Molecular Weight : 138.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart