LIF517189498 Life Chemicals Inc.

6-bromo-1,3-dihydro-2H-pyrrolo[3,2-b]pyridin-2-one

  • MDL number MFCD12963039
  • Cas Number : 1190319-62-0
  • Molecular Weight : 213.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart