LIF517189512 Life Chemicals Inc.

2-(3-chlorophenyl)-2-methylpropanenitrile

  • MDL number MFCD11036579
  • Cas Number : 64798-33-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart