LIF517189553 Life Chemicals Inc.

4-chloro-N-ethyl-6-methylpyrimidin-2-amine

  • MDL number MFCD01150900
  • Cas Number : 5748-33-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart