LIF517189555 Life Chemicals Inc.

(S)-1-(oxazol-5-yl)ethan-1-amine sesquihydrochloride

  • MDL number MFCD30179937

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart