LIF517189571 Life Chemicals Inc.

ethyl 1H,2H,3H-pyrazolo[1,5-a]imidazole-7-carboxylate

  • MDL number MFCD20684180
  • Cas Number : 1378779-72-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart